Assay Details
Gene Symbol: RDH12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500048.4 | XP_001500098.1 | 353 | 67 | ||
XM_014735664.2 | XP_014591150.1 | 355 | 67 | |||
XM_014735665.2 | XP_014591151.1 | 680 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
