Assay Details
Gene Symbol: COX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014735673.2 | XP_014591159.1 | 738 | 141 | ||
XM_023628005.1 | XP_023483773.1 | 574 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
