Assay Details
Gene Symbol: MIDEAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001916424.4 | XP_001916459.4 | 5722 | 72 | ||
XM_023628072.1 | XP_023483840.1 | 3522 | 72 | |||
XM_023628074.1 | XP_023483842.1 | 3309 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
