Assay Details
Gene Symbol: EIF2B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005605332.3 | XP_005605389.1 | 282 | 65 | ||
XM_005605333.3 | XP_005605390.1 | 214 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
