Assay Details
Gene Symbol: ACYP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491670.4 | XP_001491720.3 | 301 | 85 | ||
XM_005605334.3 | XP_005605391.2 | 252 | 85 | |||
XM_005605335.3 | XP_005605392.2 | 233 | 85 | |||
XM_023628134.1 | XP_023483902.1 | 181 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
