Assay Details
Gene Symbol: JDP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023628138.1 | XP_023483906.1 | 683 | 78 | ||
XM_023628139.1 | XP_023483907.1 | 453 | 78 | |||
XM_023628140.1 | XP_023483908.1 | 470 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
