Assay Details
Gene Symbol: ERG28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492436.6 | XP_001492486.2 | 382 | 86 | ||
XM_005605360.3 | XP_005605417.1 | 295 | 86 | |||
XM_014735734.2 | XP_014591220.1 | 382 | 86 | |||
XM_014735735.2 | XP_014591221.1 | 382 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
