Assay Details
Gene Symbol: DDX24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497967.6 | XP_001498017.2 | 2046 | 57 | ||
XM_005605419.3 | XP_005605476.2 | 2034 | 57 | |||
XM_005605420.3 | XP_005605477.2 | 2498 | 57 | |||
XM_023628325.1 | XP_023484093.1 | 2495 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
