Assay Details
Gene Symbol: SERPINA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014735763.2 | XP_014591249.2 | 1536 | 86 | ||
XM_014735764.2 | XP_014591250.2 | 1540 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
