Assay Details
Gene Symbol: GSKIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489174.5 | XP_001489224.2 | 374 | 57 | ||
XM_005605433.3 | XP_005605490.1 | 415 | 57 | |||
GenBank mRNA | JL616447.1 | - | 2 - 3 | 306 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
