Assay Details
Gene Symbol: LOC100051131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023628375.1 | XP_023484143.1 | 1153 | 68 | ||
XM_023628376.1 | XP_023484144.1 | 2206 | 68 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
