Assay Details
Gene Symbol: EIF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023628458.1 | XP_023484226.1 | 1013 | 84 | ||
XM_023628459.1 | XP_023484227.1 | 672 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
