Assay Details
Gene Symbol: TEDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023628575.1 | XP_023484343.1 | 1002 | 90 | ||
XM_023628576.1 | XP_023484344.1 | 819 | 90 | |||
XM_023628577.1 | XP_023484345.1 | 1085 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
