Assay Details
Gene Symbol: ZCCHC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504295.3 | XP_001504345.1 | 927 | 67 | ||
XM_005605629.2 | XP_005605686.1 | 925 | 67 | |||
XM_023629679.1 | XP_023485447.1 | 927 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
