Assay Details
Gene Symbol: IGFBPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023629697.1 | XP_023485465.1 | 703 | 73 | ||
XM_023629698.1 | XP_023485466.1 | 703 | 73 | |||
XM_023629699.1 | XP_023485467.1 | 703 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
