Assay Details
Gene Symbol: TMEFF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023629760.1 | XP_023485528.1 | 1201 | 80 | ||
XM_023629761.1 | XP_023485529.1 | 2348 | 80 | |||
GenBank mRNA | JL628144.1 | - | 1 - 2 | 51 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
