Assay Details
Gene Symbol: LPAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014735958.2 | XP_014591444.2 | 532 | 96 | ||
XM_023628627.1 | XP_023484395.1 | 631 | 96 | |||
XM_023628628.1 | XP_023484396.1 | 595 | 96 | |||
XM_023628629.1 | XP_023484397.1 | 465 | 96 | |||
XM_023628630.1 | XP_023484398.1 | 447 | 96 | |||
XM_023628631.1 | XP_023484399.1 | 546 | 96 | |||
XM_023628632.1 | XP_023484400.1 | 909 | 96 | |||
XM_023628633.1 | XP_023484401.1 | 381 | 96 | |||
XM_023628634.1 | XP_023484402.1 | 176 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
