Assay Details
Gene Symbol: CDC26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488609.5 | XP_001488659.3 | 327 | 105 | ||
XM_014735974.2 | XP_014591460.1 | 357 | 105 | |||
XM_023628681.1 | XP_023484449.1 | 334 | 105 | |||
GenBank mRNA | JL616342.2 | - | 1 - 2 | 83 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
