Assay Details
Gene Symbol: ARPC5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502042.5 | XP_001502092.1 | 299 | 91 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
