Assay Details
Gene Symbol: NIBAN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023628926.1 | XP_023484694.1 | 834 | 61 | ||
XM_023628927.1 | XP_023484695.1 | 694 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
