Assay Details
Gene Symbol: LCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023628982.1 | XP_023484750.1 | 341 | 57 | ||
XM_023628983.1 | XP_023484751.1 | 322 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
