Assay Details
Gene Symbol: KYAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499823.6 | XP_001499873.3 | 827 | 65 | ||
XM_005605854.3 | XP_005605911.1 | 918 | 65 | |||
XM_014736073.2 | XP_014591559.1 | 994 | 65 | |||
XM_014736074.2 | XP_014591560.1 | 772 | 65 | |||
XM_014736075.2 | XP_014591561.1 | 859 | 65 | |||
XM_014736076.2 | XP_014591562.1 | 935 | 65 | |||
XM_023629076.1 | XP_023484844.1 | 823 | 65 | |||
XM_023629077.1 | XP_023484845.1 | 721 | 65 | |||
XM_023629078.1 | XP_023484846.1 | 762 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
