Assay Details
Gene Symbol: LRRC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500261.6 | XP_001500311.1 | 124 | 98 | ||
XM_005605853.3 | XP_005605910.1 | 124 | 98 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001500261.6, XM_005605853.3 transcripts.
Set Membership: |
![]() ![]() |
