Assay Details
Gene Symbol: LRRC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005605853.3 | XP_005605910.1 | 230 | 60 | ||
XM_023629073.1 | XP_023484841.1 | 2067 | 60 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005605853.3, XM_023629073.1 transcripts.
Set Membership: |
![]() ![]() |
