Assay Details
Gene Symbol: LRRC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500261.6 | XP_001500311.1 | 2419 | 93 | ||
XM_005605853.3 | XP_005605910.1 | 2392 | 93 | |||
XM_023629073.1 | XP_023484841.1 | 4229 | 93 | |||
XM_023629074.1 | XP_023484842.1 | 2258 | 93 | |||
XM_023629075.1 | XP_023484843.1 | 2245 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
