Assay Details
Gene Symbol: TARBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504534.5 | XP_001504584.1 | 1287 | 54 | ||
XM_001504538.5 | XP_001504588.1 | 1067 | 54 | |||
XM_005611249.3 | XP_005611306.1 | 1283 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
