Assay Details
Gene Symbol: NAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488470.6 | XP_001488520.2 | 1405 | 63 | ||
XM_005611328.3 | XP_005611385.1 | 1406 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
