Assay Details
Gene Symbol: MARS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488891.5 | XP_001488941.1 | 1289 | 62 | ||
XM_005611342.3 | XP_005611399.1 | 1289 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
