Assay Details
Gene Symbol: PIP4K2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023643877.1 | XP_023499645.1 | 1267 | 67 | ||
XM_023643878.1 | XP_023499646.1 | 1267 | 67 | |||
XM_023643879.1 | XP_023499647.1 | 1228 | 67 | |||
XM_023643880.1 | XP_023499648.1 | 1154 | 67 | |||
XM_023643881.1 | XP_023499649.1 | 1241 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
