Assay Details
Gene Symbol: AIF1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023629157.1 | XP_023484925.1 | 410 | 65 | ||
XM_023629158.1 | XP_023484926.1 | 287 | 65 | |||
XM_023629159.1 | XP_023484927.1 | 1146 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
