Assay Details
Gene Symbol: AIF1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023629157.1 | XP_023484925.1 | 450 | 97 | ||
XM_023629158.1 | XP_023484926.1 | 327 | 97 | |||
XM_023629159.1 | XP_023484927.1 | 1186 | 97 |
