Assay Details
Gene Symbol: MED27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498768.4 | XP_001498818.1 | 611 | 62 | ||
XM_023629190.1 | XP_023484958.1 | 611 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
