Assay Details
Gene Symbol: SPACA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005605900.3 | XP_005605957.1 | 335 | 107 | ||
XM_014736103.2 | XP_014591589.1 | 336 | 107 | |||
XM_014736104.2 | XP_014591590.1 | 300 | 107 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005605900.3, XM_014736103.2, XM_014736104.2 transcripts.
Set Membership: |
![]() ![]() |
