Assay Details
Gene Symbol: SPACA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005605900.3 | XP_005605957.1 | 513 | 62 | ||
XM_014736103.2 | XP_014591589.1 | 514 | 62 | |||
XM_014736104.2 | XP_014591590.1 | 478 | 62 | |||
XM_014736106.2 | XP_014591592.1 | 330 | 62 | |||
XM_014736107.2 | XP_014591593.1 | 361 | 62 | |||
XM_014736108.2 | XP_014591594.1 | 292 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
