Assay Details
Gene Symbol: AGPAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023629309.1 | XP_023485077.1 | 713 | 105 | ||
XM_023629310.1 | XP_023485078.1 | 685 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
