Assay Details
Gene Symbol: CCDC187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023629344.1 | XP_023485112.1 | 3995 | 104 | ||
XM_023629345.1 | XP_023485113.1 | 2929 | 104 | |||
XM_023629346.1 | XP_023485114.1 | 3010 | 104 | |||
XM_023629347.1 | XP_023485115.1 | 3493 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
