Assay Details
Gene Symbol: ENTPD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490836.4 | XP_001490886.1 | 1303 | 48 | ||
XM_014736172.2 | XP_014591658.1 | 1303 | 48 | |||
XM_023629494.1 | XP_023485262.1 | 4260 | 48 | |||
XM_023629495.1 | XP_023485263.1 | 982 | 48 | |||
XM_023629496.1 | XP_023485264.1 | 1303 | 48 | |||
XM_023629497.1 | XP_023485265.1 | 1303 | 48 | |||
XM_023629498.1 | XP_023485266.1 | 1303 | 48 | |||
XM_023629499.1 | XP_023485267.1 | 1303 | 48 |

More Information
Set Membership: |
![]() ![]() ![]() |
