Assay Details
Gene Symbol: CXADR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023629904.1 | XP_023485672.1 | 778 | 59 | ||
XM_023629905.1 | XP_023485673.1 | 778 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
