Assay Details
Gene Symbol: IFNGR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023630057.1 | XP_023485825.1 | 1183 | 62 | ||
GenBank mRNA | JL625602.1 | - | 3 - 4 | 1006 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
