Assay Details
Gene Symbol: RCAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003364201.4 | XP_003364249.1 | 450 | 79 | ||
XM_014736353.2 | XP_014591839.2 | 577 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
