Assay Details
Gene Symbol: CLIC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493952.4 | XP_001494002.2 | 1645 | 110 | ||
XM_023630089.1 | XP_023485857.1 | 2050 | 110 |

More Information
Set Membership: |
![]() ![]() |
