Assay Details
Gene Symbol: DOP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023630119.1 | XP_023485887.1 | 879 | 57 | ||
XM_023630120.1 | XP_023485888.1 | 879 | 57 | |||
XM_023630121.1 | XP_023485889.1 | 879 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
