Assay Details
Gene Symbol: ERG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494781.5 | XP_001494831.1 | 1038 | 77 | ||
XM_003364202.4 | XP_003364250.1 | 1255 | 77 | |||
XM_005606152.3 | XP_005606209.1 | 1184 | 77 | |||
XM_023630154.1 | XP_023485922.1 | 1185 | 77 | |||
XM_023630155.1 | XP_023485923.1 | 1130 | 77 | |||
XM_023630156.1 | XP_023485924.1 | 1150 | 77 | |||
XM_023630157.1 | XP_023485925.1 | 1069 | 77 | |||
XM_023630158.1 | XP_023485926.1 | 1401 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
