Assay Details
Gene Symbol: SH3BGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005606156.3 | XP_005606213.1 | 535 | 81 | ||
XM_023630192.1 | XP_023485960.1 | 532 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
