Assay Details
Gene Symbol: RIPK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023630208.1 | XP_023485976.1 | 680 | 57 | ||
XM_023630209.1 | XP_023485977.1 | 679 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
