Assay Details
Gene Symbol: U2AF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005606173.3 | XP_005606230.1 | 371 | 72 | ||
XM_023630243.1 | XP_023486011.1 | 594 | 72 | |||
XM_023630244.1 | XP_023486012.1 | 594 | 72 | |||
XM_023630245.1 | XP_023486013.1 | 601 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
