Assay Details
Gene Symbol: UBE2G2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490058.6 | XP_001490108.4 | 667 | 60 | ||
XM_005606180.3 | XP_005606237.1 | 361 | 60 | |||
XM_023630294.1 | XP_023486062.1 | 488 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
