Assay Details
Gene Symbol: SMIM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488864.6 | XP_001488914.2 | 539 | 68 | ||
XM_005606270.3 | XP_005606327.1 | 535 | 68 | |||
XM_023630902.1 | XP_023486670.1 | 526 | 68 | |||
XM_023630903.1 | XP_023486671.1 | 506 | 68 | |||
XM_023630905.1 | XP_023486673.1 | 613 | 68 | |||
XM_023630906.1 | XP_023486674.1 | 593 | 68 | |||
XM_023630907.1 | XP_023486675.1 | 363 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
