Assay Details
Gene Symbol: GOLGA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490279.6 | XP_001490329.1 | 250 | 111 | ||
XM_005606286.3 | XP_005606343.1 | 257 | 111 | |||
XM_014736537.2 | XP_014592023.1 | 250 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
