Assay Details
Gene Symbol: VPS37A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488521.5 | XP_001488571.1 | 893 | 64 | ||
GenBank mRNA | JL616412.1 | - | 4 - 5 | 503 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
